Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brdt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054054.2 | NP_473395.2 | 10 - 11 | 1757 | 72 | |
GenBank mRNA | AF358660.1 | - | 10 - 11 | 1792 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |