Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brdt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054054.2 | NP_473395.2 | 7 - 8 | 1270 | 116 | |
GenBank mRNA | AF358660.1 | - | 7 - 8 | 1305 | 116 | |
BY726053.1 | - | 3 - 4 | 596 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |