Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080798.1 | NP_001074267.1 | 11 - 12 | 2623 | 80 | |
NM_133919.3 | NP_598680.3 | 11 - 12 | 2634 | 80 | ||
GenBank mRNA | AF013131.1 | - | 10 - 11 | 2449 | 80 | |
AK139900.1 | - | 3 - 4 | 1311 | 80 | ||
AK143742.1 | - | 11 - 12 | 2623 | 80 | ||
AY749168.1 | - | 11 - 12 | 2467 | 80 | ||
BC131665.1 | - | 11 - 12 | 2467 | 40110795 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |