Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080798.1 | NP_001074267.1 | 14 - 15 | 3063 | 81 | |
NM_133919.3 | NP_598680.3 | 14 - 15 | 3074 | 81 | ||
GenBank mRNA | AF013131.1 | - | 13 - 14 | 2889 | 81 | |
AF074266.1 | - | 14 - 15 | 2904 | 81 | ||
AY749168.1 | - | 14 - 15 | 2907 | 81 | ||
BC131665.1 | - | 14 - 15 | 2907 | 40110795 | 81 | |
CN537099.1 | - | 1 - 2 | 72 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |