Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199677.1 | NP_001186606.1 | 3 - 4 | 462 | 82 | |
GenBank mRNA | AK158287.1 | - | 3 - 4 | 391 | 82 | |
AY081953.2 | - | 3 - 4 | 462 | 82 | ||
BC044789.1 | - | 3 - 4 | 365 | 3484221 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |