Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025644.4 | NP_079920.1 | 3 - 4 | 248 | 68 | |
GenBank mRNA | AK006018.1 | - | 3 - 4 | 236 | 68 | |
AK011817.1 | - | 3 - 4 | 231 | 68 | ||
BC024423.1 | - | 3 - 4 | 228 | 5010850 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |