Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199676.1 | NP_001186605.1 | 10 - 11 | 1326 | 57 | |
NM_145358.2 | NP_663333.1 | 10 - 11 | 1326 | 57 | ||
GenBank mRNA | AF453383.1 | - | 9 - 10 | 1109 | 57 | |
AK031399.1 | - | 10 - 11 | 1251 | 57 | ||
AK032070.1 | - | 10 - 11 | 1299 | 57 | ||
AK044660.1 | - | 10 - 11 | 1315 | 57 | ||
AK122370.1 | - | 5 - 6 | 492 | 57 | ||
BC023103.1 | - | 9 - 10 | 1159 | 3993474 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |