Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198306.3 | NP_938048.2 | 6 - 7 | 1428 | 59 | |
GenBank mRNA | AK032568.1 | - | 6 - 7 | 1431 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |