Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289589.1 | NP_001276518.1 | 1 - 2 | 222 | 125 | |
NM_001289590.1 | NP_001276519.1 | 1 - 2 | 222 | 125 | ||
NM_177681.4 | NP_808349.1 | 1 - 2 | 222 | 125 | ||
GenBank mRNA | AK049650.1 | - | 1 - 2 | 178 | 125 | |
BB612291.1 | - | 1 - 2 | 222 | 125 | ||
BC053080.1 | - | 1 - 2 | 131 | 5702966 | 125 | |
BC059836.1 | - | 1 - 2 | 133 | 5697949 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |