Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actl6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031404.4 | NP_113581.1 | 4 - 5 | 453 | 104 | |
GenBank mRNA | AB075226.1 | - | 4 - 5 | 373 | 104 | |
BC049539.1 | - | 4 - 5 | 453 | 6534039 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |