Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289511.1 | NP_001276440.1 | 1 - 2 | 60 | 70 | |
GenBank mRNA | AF222895.1 | - | 1 - 2 | 60 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |