Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rph3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB112924.1 | - | 9 - 10 | 892 | 93 | |
AK139040.1 | - | 8 - 9 | 834 | 93 | ||
BC042585.1 | - | 9 - 10 | 1059 | 5364086 | 93 | |
BC050883.1 | - | 9 - 10 | 1026 | 5698459 | 93 | |
D29965.1 | - | 9 - 10 | 869 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |