Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001446.3 | NP_001001446.3 | 1 - 2 | 243 | 105 | |
NM_001167905.1 | NP_001161377.1 | 1 - 2 | 243 | 105 | ||
GenBank mRNA | AK050436.1 | - | 1 - 2 | 205 | 105 | |
AK149451.1 | - | 1 - 2 | 205 | 105 | ||
BC022141.1 | - | 1 - 2 | 193 | 105 | ||
BC024068.1 | - | 1 - 2 | 163 | 105 | ||
BC025819.1 | - | 1 - 2 | 195 | 5099385 | 105 | |
BC026766.1 | - | 1 - 2 | 195 | 4195655 | 105 | |
BC034834.1 | - | 1 - 2 | 188 | 5099935 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |