Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025307.1 | NP_001020478.1 | 2 - 3 | 803 | 67 | |
NM_001286543.1 | NP_001273472.1 | 2 - 3 | 803 | 67 | ||
NM_152220.2 | NP_689344.1 | 2 - 3 | 803 | 67 | ||
GenBank mRNA | AK038415.1 | - | 2 - 3 | 310 | 67 | |
AK170105.1 | - | 2 - 3 | 212 | 67 | ||
AK171135.1 | - | 2 - 3 | 346 | 67 | ||
BC024844.1 | - | 2 - 3 | 211 | 5357204 | 67 | |
BC056949.1 | - | 2 - 3 | 803 | 67 | ||
D29797.1 | - | 2 - 3 | 224 | 67 | ||
D29798.1 | - | 2 - 3 | 222 | 67 | ||
D29800.1 | - | 2 - 3 | 135 | 67 | ||
D38375.1 | - | 2 - 3 | 121 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |