Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008926.4 | NP_032952.3 | 1 - 2 | 156 | 63 | |
GenBank mRNA | AK038739.1 | - | 1 - 2 | 140 | 63 | |
AK081422.1 | - | 1 - 2 | 157 | 63 | ||
BC113205.1 | - | 1 - 2 | 67 | 40047751 | 63 | |
L12460.1 | - | 1 - 2 | 428 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |