Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201470.1 | NP_001188399.1 | 2 - 3 | 278 | 89 | |
NM_011864.3 | NP_035994.2 | 2 - 3 | 299 | 89 | ||
GenBank mRNA | AF052453.1 | - | 2 - 3 | 280 | 89 | |
AF085144.1 | - | 2 - 3 | 210 | 89 | ||
AK128962.1 | - | 2 - 3 | 299 | 89 | ||
AK137675.1 | - | 2 - 3 | 204 | 89 | ||
BC090997.1 | - | 2 - 3 | 299 | 30121331 | 89 | |
BY224664.1 | - | 2 - 3 | 279 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |