Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031376.3 | NP_113553.1 | 6 - 7 | 1098 | 83 | |
GenBank mRNA | AF293806.1 | - | 6 - 7 | 1098 | 83 | |
AK087722.1 | - | 4 - 5 | 567 | 83 | ||
AK089785.1 | - | 6 - 7 | 1131 | 83 | ||
AK150114.1 | - | 4 - 5 | 567 | 83 | ||
AK153548.1 | - | 4 - 5 | 549 | 83 | ||
AK154326.1 | - | 6 - 7 | 1126 | 83 | ||
AK170130.1 | - | 6 - 7 | 1131 | 83 | ||
AK170925.1 | - | 6 - 7 | 1131 | 83 | ||
AK171021.1 | - | 6 - 7 | 1126 | 83 | ||
AK172670.1 | - | 3 - 4 | 1550 | 83 | ||
BC113141.1 | - | 6 - 7 | 1057 | 40047744 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |