Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167921.1 | NP_001161393.1 | 3 - 4 | 343 | 107 | |
NM_001167922.1 | NP_001161394.1 | 3 - 4 | 445 | 107 | ||
NM_026584.3 | NP_080860.3 | 3 - 4 | 331 | 107 | ||
GenBank mRNA | AK007492.1 | - | 2 - 3 | 288 | 107 | |
AK021183.1 | - | 3 - 4 | 296 | 107 | ||
AK088342.1 | - | 3 - 4 | 356 | 107 | ||
AK146803.1 | - | 3 - 4 | 318 | 107 | ||
AK150415.1 | - | 3 - 4 | 289 | 107 | ||
AK153370.1 | - | 3 - 4 | 289 | 107 | ||
AK158783.1 | - | 3 - 4 | 425 | 107 | ||
BC020016.1 | - | 3 - 4 | 311 | 3501113 | 107 | |
CX239577.1 | - | 3 - 4 | 343 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |