Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sqle | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009270.3 | NP_033296.1 | 7 - 8 | 1955 | 91 | |
GenBank mRNA | AK148419.1 | - | 7 - 8 | 1940 | 91 | |
AK163502.1 | - | 7 - 8 | 1935 | 91 | ||
AK180804.1 | - | 2 - 3 | 91 | |||
AK189403.1 | - | 1 - 2 | 43 | 91 | ||
AK208956.1 | - | 1 - 2 | 43 | 91 | ||
AK209538.1 | - | 1 - 2 | 43 | 91 | ||
BC042781.1 | - | 7 - 8 | 1212 | 4915911 | 91 | |
BC056361.1 | - | 7 - 8 | 1955 | 6816800 | 91 | |
D42048.1 | - | 7 - 8 | 1821 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |