Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109992.1 | NP_001103462.1 | 8 - 9 | 1118 | 80 | |
NM_011202.3 | NP_035332.1 | 8 - 9 | 1118 | 80 | ||
GenBank mRNA | AK147354.1 | - | 7 - 8 | 2964 | 80 | |
AK159501.1 | - | 8 - 9 | 1118 | 80 | ||
AK159587.1 | - | 8 - 9 | 1118 | 80 | ||
AK163809.1 | - | 5 - 6 | 546 | 80 | ||
BC057398.1 | - | 8 - 9 | 1050 | 6409640 | 80 | |
BC059278.1 | - | 8 - 9 | 1096 | 2866038 | 80 | |
D84372.1 | - | 8 - 9 | 994 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |