Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172716.4 | NP_766304.2 | 8 - 9 | 767 | 72 | |
GenBank mRNA | AK089091.1 | - | 8 - 9 | 768 | 72 | |
BC049266.1 | - | 634 | 5371040 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |