Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181400.3 | NP_852065.2 | 3 - 4 | 566 | 135 | |
GenBank mRNA | AK047357.1 | - | 3 - 4 | 566 | 135 | |
AK163422.1 | - | 3 - 4 | 457 | 135 | ||
BC040337.1 | - | 3 - 4 | 427 | 3586908 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |