Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181410.3 | NP_852075.1 | 10 - 11 | 730 | 72 | |
GenBank mRNA | AK050227.1 | - | 10 - 11 | 715 | 72 | |
AK133941.1 | - | 10 - 11 | 701 | 72 | ||
BC017515.1 | - | 10 - 11 | 707 | 4504077 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |