Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007941.3 | NP_031967.2 | 10 - 11 | 1059 | 103 | |
GenBank mRNA | AK146462.1 | - | 10 - 11 | 988 | 103 | |
BC046279.1 | - | 10 - 11 | 983 | 6392250 | 103 | |
D10475.1 | - | 10 - 11 | 1062 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |