Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113529.1 | NP_001107001.1 | 5 - 6 | 945 | 99 | |
GenBank mRNA | BC025593.1 | - | 5 - 6 | 731 | 99 | |
S78392.1 | - | 5 - 6 | 698 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |