Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163350.1 | NP_001156822.1 | 4 - 5 | 2155 | 132 | |
GenBank mRNA | AB038664.1 | - | 3 - 4 | 1060 | 132 | |
AF475072.1 | - | 3 - 4 | 1060 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |