Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284409.1 | NP_001271338.1 | 1 - 2 | 211 | 83 | |
GenBank mRNA | AK014231.1 | - | 1 - 2 | 96 | 83 | |
AK146226.1 | - | 1 - 2 | 212 | 83 | ||
BC038825.1 | - | 1 - 2 | 95 | 4482368 | 83 | |
Y13086.1 | - | 1 - 2 | 52 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons |