Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008975.3 | NP_033001.2 | 1 - 2 | 441 | 60 | |
GenBank mRNA | AK014601.1 | - | 1 - 2 | 56 | 60 | |
AK143702.1 | - | 1 - 2 | 93 | 60 | ||
AK172192.1 | - | 1 - 2 | 95 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mus musculus 1700 Probe spans exons |