Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp11b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009991.3 | NP_034121.3 | 3 - 4 | 690 | 66 | |
GenBank mRNA | BC116908.1 | - | 3 - 4 | 690 | 40126227 | 66 |
BC119321.1 | - | 3 - 4 | 690 | 8734070 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |