Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005863.2 | NP_001005863.1 | 7 - 8 | 3117 | 75 | |
NM_001005864.3 | NP_001005864.1 | 5 - 6 | 723 | 75 | ||
NM_001005865.3 | NP_001005865.2 | 3 - 4 | 736 | 75 | ||
NM_001286413.1 | NP_001273342.1 | 7 - 8 | 2999 | 75 | ||
GenBank mRNA | AF173380.1 | - | 3 - 4 | 575 | 75 | |
AF493235.1 | - | 3 - 4 | 640 | 75 | ||
AK030510.1 | - | 3 - 4 | 700 | 75 | ||
AK031693.1 | - | 3 - 4 | 670 | 75 | ||
AK035576.1 | - | 1 - 2 | 104 | 75 | ||
AK129324.1 | - | 3 - 4 | 722 | 75 | ||
AY246699.1 | - | 3 - 4 | 398 | 75 | ||
AY626781.1 | - | 3 - 4 | 733 | 75 | ||
AY626782.1 | - | 5 - 6 | 721 | 75 | ||
AY626783.1 | - | 6 - 7 | 2939 | 75 | ||
BC041777.1 | - | 6 - 7 | 1782 | 4240274 | 75 | |
BC042206.1 | - | 6 - 7 | 1782 | 4240274 | 75 | |
BC043321.1 | - | 5 - 6 | 687 | 75 | ||
BC089009.1 | - | 7 - 8 | 2999 | 30543240 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |