Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sharpin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025340.2 | NP_079616.2 | 7 - 8 | 1457 | 57 | |
GenBank mRNA | AB052763.1 | - | 7 - 8 | 1443 | 57 | |
AK002878.1 | - | 7 - 8 | 1028 | 57 | ||
AK003578.1 | - | 7 - 8 | 1037 | 57 | ||
AK165824.1 | - | 4 - 5 | 1384 | 57 | ||
AK171774.1 | - | 6 - 7 | 1631 | 57 | ||
AK182331.1 | - | 1 - 2 | 83 | 57 | ||
AK183724.1 | - | 1 - 2 | 83 | 57 | ||
AK188032.1 | - | 1 - 2 | 83 | 57 | ||
BC016203.1 | - | 7 - 8 | 1398 | 4527839 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |