Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256096.1 | NP_001243025.1 | 7 - 8 | 1628 | 68 | |
NM_001256097.1 | NP_001243026.1 | 7 - 8 | 1568 | 68 | ||
NM_001256098.1 | NP_001243027.1 | 6 - 7 | 1490 | 68 | ||
NM_023742.2 | NP_076231.1 | 7 - 8 | 1625 | 68 | ||
GenBank mRNA | AB015423.1 | - | 7 - 8 | 1548 | 68 | |
AB015424.1 | - | 6 - 7 | 1413 | 68 | ||
AK012137.1 | - | 7 - 8 | 1624 | 68 | ||
AK044105.1 | - | 7 - 8 | 1525 | 68 | ||
AK054443.1 | - | 6 - 7 | 1480 | 68 | ||
AK173192.1 | - | 2 - 3 | 4406 | 68 | ||
AK185107.1 | - | 1 - 2 | 51 | 68 | ||
BC024925.1 | - | 8 - 9 | 1645 | 3711019 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |