Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039162.2 | NP_001034251.1 | 1 - 2 | 389 | 99 | |
NM_009990.3 | NP_034120.2 | 1 - 2 | 389 | 99 | ||
GenBank mRNA | AK045967.1 | - | 1 - 2 | 232 | 99 | |
AK172928.1 | - | 1 - 2 | 282 | 99 | ||
BC039162.1 | - | 1 - 2 | 329 | 3498067 | 99 | |
BC053048.1 | - | 1 - 2 | 389 | 5709279 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |