Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256096.1 | NP_001243025.1 | 6 - 7 | 1553 | 105 | |
GenBank mRNA | AK012137.1 | - | 6 - 7 | 1549 | 105 | |
BC024925.1 | - | 7 - 8 | 1570 | 3711019 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |