Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evi5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007964.2 | NP_031990.2 | 3 - 4 | 540 | 89 | |
GenBank mRNA | AK134294.1 | - | 3 - 4 | 741 | 89 | |
AK165332.1 | - | 4 - 5 | 533 | 89 | ||
AK165885.1 | - | 3 - 4 | 493 | 89 | ||
BC030056.1 | - | 4 - 5 | 547 | 89 | ||
BI737814.1 | - | 3 - 4 | 464 | 89 | ||
U53586.1 | - | 3 - 4 | 540 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |