Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mad1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010752.3 | NP_034882.2 | 1 - 2 | 44 | 68 | |
GenBank mRNA | AF083812.1 | - | 1 - 2 | 51 | 68 | |
AF083813.1 | - | 1 - 2 | 51 | 68 | ||
AK136981.1 | - | 1 - 2 | 46 | 68 | ||
BC010702.1 | - | 1 - 2 | 42 | 3156639 | 68 | |
BI735882.1 | - | 1 - 2 | 44 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |