Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172412.2 | NP_766000.1 | 4 - 5 | 853 | 65 | |
GenBank mRNA | AK010515.1 | - | 4 - 5 | 883 | 65 | |
AK049639.1 | - | 4 - 5 | 854 | 65 | ||
AK160959.1 | - | 4 - 5 | 899 | 65 | ||
BC083180.1 | - | 4 - 5 | 860 | 5707617 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |