Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194940.2 | NP_001181869.1 | 13 - 14 | 4227 | 74 | |
NM_001194941.1 | NP_001181870.1 | 9 - 10 | 2857 | 74 | ||
NM_015802.3 | NP_056617.2 | 9 - 10 | 2775 | 74 | ||
GenBank mRNA | AF178078.1 | - | 9 - 10 | 2711 | 74 | |
AK147539.1 | - | 9 - 10 | 2857 | 74 | ||
AK162413.1 | - | 9 - 10 | 2776 | 74 | ||
AK166175.1 | - | 9 - 10 | 2735 | 74 | ||
BC150802.1 | - | 9 - 10 | 2560 | 9087713 | 74 | |
HM008381.1 | - | 12 - 13 | 3780 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |