Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276287.1 | NP_001263216.1 | 11 - 12 | 1681 | 57 | |
NM_181066.3 | NP_851411.1 | 11 - 12 | 1583 | 57 | ||
NR_074085.1 | 10 - 11 | 1491 | 57 | |||
GenBank mRNA | AF226663.1 | - | 1 - 2 | 130 | 57 | |
AK049321.1 | - | 10 - 11 | 1480 | 57 | ||
AK084987.1 | - | 11 - 12 | 1572 | 57 | ||
AK085091.1 | - | 11 - 12 | 1572 | 57 | ||
AK154448.1 | - | 11 - 12 | 1554 | 57 | ||
AK170269.1 | - | 11 - 12 | 1577 | 57 | ||
BC068301.1 | - | 11 - 12 | 1647 | 30061848 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |