Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080808.1 | NP_001074277.1 | 7 - 8 | 1447 | 80 | |
GenBank mRNA | BC024804.1 | - | 7 - 8 | 1440 | 80 | |
BC126948.1 | - | 7 - 8 | 1447 | 40090576 | 80 | |
GU973876.1 | - | 7 - 8 | 1447 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |