Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173405.2 | NP_775581.1 | 6 - 7 | 1130 | 56 | |
GenBank mRNA | AJ879912.1 | - | 4 - 5 | 771 | 56 | |
AK078332.1 | - | 6 - 7 | 1130 | 56 | ||
AK155136.1 | - | 1017 | 56 | |||
AK170176.1 | - | 5 - 6 | 1104 | 56 | ||
BC046428.1 | - | 1 - 2 | 98 | 56 | ||
BC129841.1 | - | 4 - 5 | 913 | 40090727 | 56 | |
BC129842.1 | - | 4 - 5 | 865 | 40090729 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |