Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vkorc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001327.2 | NP_001001327.1 | 1 - 2 | 72 | ||
GenBank mRNA | AK078942.1 | - | 1 - 2 | 72 | ||
BB625262.1 | - | 1 - 2 | 72 | |||
BC085133.1 | - | 1 - 2 | 6733526 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |