Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145227.3 | NP_660262.2 | 5 - 6 | 1195 | 65 | |
GenBank mRNA | AB067535.1 | - | 5 - 6 | 1200 | 65 | |
AF418010.1 | - | 5 - 6 | 1135 | 65 | ||
BC118538.1 | - | 5 - 6 | 1135 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |