Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgef1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145839.2 | NP_665838.1 | 8 - 9 | 1145 | 81 | |
NM_181318.4 | NP_851835.1 | 8 - 9 | 1142 | 81 | ||
GenBank mRNA | AK028756.1 | - | 8 - 9 | 1034 | 81 | |
AK090085.1 | - | 8 - 9 | 1037 | 81 | ||
AK149812.1 | - | 8 - 9 | 1064 | 81 | ||
AY129963.1 | - | 8 - 9 | 1072 | 81 | ||
BC050858.1 | - | 8 - 9 | 1075 | 6517502 | 81 | |
BC052726.1 | - | 8 - 9 | 1036 | 6813357 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |