Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168525.1 | NP_001161997.1 | 5 - 6 | 969 | 75 | |
NM_144792.4 | NP_659041.3 | 5 - 6 | 537 | 75 | ||
GenBank mRNA | AK076554.1 | - | 3 - 4 | 202 | 75 | |
AK082974.1 | - | 5 - 6 | 466 | 75 | ||
AK146782.1 | - | 5 - 6 | 537 | 75 | ||
AK151170.1 | - | 5 - 6 | 522 | 75 | ||
AK153406.1 | - | 5 - 6 | 522 | 75 | ||
AY509044.1 | - | 5 - 6 | 537 | 75 | ||
BC076618.1 | - | 5 - 6 | 969 | 75 | ||
BC085298.1 | - | 2 - 3 | 131 | 6307174 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |