Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK076554.1 | - | 5 - 6 | 1088 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |