Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024253.4 | NP_077215.2 | 1 - 2 | 322 | 108 | |
GenBank mRNA | AK010872.1 | - | 1 - 2 | 322 | 108 | |
AK088383.1 | - | 1 - 2 | 331 | 108 | ||
BC055882.1 | - | 1 - 2 | 387 | 3497559 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |