Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps18c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026826.1 | NP_081102.1 | 3 - 4 | 244 | 90 | |
GenBank mRNA | AK004139.1 | - | 3 - 4 | 257 | 90 | |
AK088290.1 | - | 3 - 4 | 257 | 90 | ||
AK145935.1 | - | 3 - 4 | 261 | 90 | ||
BC028442.1 | - | 3 - 4 | 244 | 1196057 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |