Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178741.3 | NP_848856.1 | 4 - 5 | 1234 | 135 | |
GenBank mRNA | AK031028.1 | - | 4 - 5 | 1235 | 135 | |
AK034618.1 | - | 4 - 5 | 1200 | 135 | ||
AK049960.1 | - | 3 - 4 | 1122 | 135 | ||
AY101769.1 | - | 3 - 4 | 979 | 135 | ||
BC086802.1 | - | 3 - 4 | 1095 | 30612176 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |