Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025573.3 | NP_079849.1 | 2 - 3 | 484 | 60 | |
GenBank mRNA | AK011621.1 | - | 2 - 3 | 444 | 60 | |
AK020084.1 | - | 2 - 3 | 256 | 60 | ||
AK159190.1 | - | 2 - 3 | 482 | 60 | ||
BC012217.1 | - | 2 - 3 | 484 | 3483731 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |